CAS RN 842148-40-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AKT Kinase Inhibitor, MedChemExpress
MedChemExpress AKT Kinase Inhibitor is an Akt kinase inhibitor with anti-tumor activity.