CAS RN 836620-48-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ AS 1842856
Potent and selective Foxo1 inhibitor; orally bioavailable
2
AS1842856, MedChemExpress
MedChemExpress AS1842856, a specific Foxo1 inhibitor (IC50=30 nM), potently suppresses autophagy. AS1842856 only reduces the activity of FoxO1 by binding with it, without affecting its transcription and protein expression.