CAS RN 833481-73-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
2OH-BNPP1, MedChemExpress
MedChemExpress 2OH-BNPP1 is an inhibitor of BUB1 kinase, a Ser/Thr kinase, used for the treatment of cancer.