CAS RN 82373-94-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
2,3,5,4'-Tetrahydroxystilbene 2-O-β-D-glucoside, MedChemExpress
MedChemExpress 2,3,4',5-tetrahydroxystilbene 2-O-D-glucoside isolats from the roots of Polygonum species, inhibits the formation of 5-HETE, HHT and thromboxane B2, although less strongly.