CAS RN 819812-04-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
KW-2478, MedChemExpress
MedChemExpress KW-2478 is an inhibitor of Hsp90α, with an IC50 of 3.8 nM, and has antitumor activity against various human hematological tumor cells.