CAS RN 815610-63-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Ustekinumab
MedChemExpress Ustekinumab is an anti-IL-12/IL-23 IgG1κ human monoclonal antibody.
2
MedChemExpress Ustekinumab
MedChemExpress Ustekinumab is an anti-IL-12/IL-23 IgG1κ human monoclonal antibody.