CAS RN 80890-47-7
IUPAC Nombre:
(2R,3S,4R,6R)-6-{[(2R,4R,5S,6R)-2-[(2S,3R,4S)-4-[(2R,3S,4E,6E,9R,10S,11S,12R,13R,14E,16Z)-11-ethyl-10,12-dihydroxy-3,17-dimethoxy-7,9,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6,14,16-tetraen-2-yl]-3-hydroxypentan-2-yl]-2-hydroxy-5-methyl-6-[(1E)-prop-1-en-1-yl]oxan-4-yl]oxy}-4-hydroxy-2-methyloxan-3-yl carbamate
Sinónimos:
(2R,3S,4R,6R)-6-{[(2R,4R,5S,6R)-2-[(2S,3R,4S)-4-[(2R,3S,4E,6E,9R,10S,11S,12R,13R,14E,16Z)-11-ethyl-10,12-dihydroxy-3,17-dimethoxy-7,9,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6,14,16-tetraen-2-yl]-3-hydroxypentan-2-yl]-2-hydroxy-5-methyl-6-[(1E)-prop-1-en-1-yl]oxan-4-yl]oxy}-4-hydroxy-2-methyloxan-3-yl carbamate
Peso molecular (g/mol):
866.1
Fórmula molecular:
C46H75NO14
InChi Key:
DJZCTUVALDDONK-HQMSUKCRSA-N
SMILES:
CC[C@H]1[C@@H](O)[C@H](C)C\C(C)=C\C=C\[C@H](OC)[C@H](OC(=O)\C(OC)=C\C(\C)=C\[C@@H](C)[C@H]1O)[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](O[C@H]2C[C@@H](O)[C@H](OC(N)=O)[C@@H](C)O2)[C@H](C)[C@H](O1)\C=C\C
1
–
2
de
2
resultados
1
Concanamycin A, MedChemExpress
MedChemExpress Concanamycin A (Folimycin; Antibiotic X 4357B) is a macrolide antibiotic, a vacuolar type H+-ATPase (V-ATPase) inhibitor. Concanamycin A is also an inhibitor of lysosomal acidification, can be used to T cell-mediated inflammation research-.