CAS RN 803647-40-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NSC59984, MedChemExpress
MedChemExpress NSC59984 induces mutant p53 protein degradation via MDM2 and the ubiquitin-proteasome pathway. NSC59984 acts by targeting GOF-mutant p53 and stimulates p73 to restore the p53 pathway signaling.