CAS RN 794568-92-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BAY 73-6691, MedChemExpress
MedChemExpress BAY 73-6691 ((R)-BAY 73-6691) is a potent, brain penetrant, and selective PDE9A inhibitor.