CAS RN 79243-10-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Somatostatin-28 (1-14)
MedChemExpress Somatostatin-28 (1-14) is an N-terminal fragment of the neuropeptide somatostatin-28.
2
MedChemExpress Somatostatin-28 (1-14)
MedChemExpress Somatostatin-28 (1-14) is an N-terminal fragment of the neuropeptide somatostatin-28.
3
MedChemExpress Somatostatin-28 (1-14)
MedChemExpress Somatostatin-28 (1-14) is an N-terminal fragment of the neuropeptide somatostatin-28.