CAS RN 792236-07-8
IUPAC Nombre:
(methylcarbamoyl)methyl N-{2-[1-(6-methylpyridin-2-yl)piperidin-4-yl]ethyl}carbamate
Sinónimos:
(methylcarbamoyl)methyl N-{2-[1-(6-methylpyridin-2-yl)piperidin-4-yl]ethyl}carbamate
Peso molecular (g/mol):
334.42
Fórmula molecular:
C17H26N4O3
InChi Key:
HSYCMGWPPRTNKH-UHFFFAOYSA-N
SMILES:
CNC(=O)COC(=O)NCCC1CCN(CC1)C1=CC=CC(C)=N1
1
–
1
de
1
resultados
1
SA 47, MedChemExpress
MedChemExpress SA 47 is a selective and potent inhibitor of fatty acid amide hydrolase (FAAH) and carbamate.