CAS RN 790702-57-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ NGI 1
Oligosaccharyltransferase (OST) inhibitor; anti-flaviviral
2
NGI-1, MedChemExpress
MedChemExpress NGI-1 (ML414) is a potent oligosaccharyltransferase (OST) inhibitor, directly targeting and blocking the function of the OST catalytic subunits STT3A and STT3B. NGI-1 is a cell permeable inhibitor and can effectively reduce virus infectivity without affecting cell viability.