CAS RN 79-61-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Dichlorisone acetate, MedChemExpress
MedChemExpress Dichlorisone acetate is a synthetic glucocorticoid corticosteroid used as an anti-inflammatory agent.