CAS RN 78950-78-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
8-OH-DPAT, MedChemExpress
MedChemExpress 8-OH-DPAT is a potent and selective 5-HT agonist, with a pIC50 of 8.19 for 5-HT1A and a Ki of 466 nM for 5-HT7; 8-OH-DPAT weakly binds to 5-HT1B (pIC50, 5.42), 5-HT (pIC50 <5).