CAS RN 786643-20-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress D149 Dye
MedChemExpress D149 Dye is an indoline-based dye, which is a high-extinction-coefficient metal-free organic sensitizer.