CAS RN 78281-02-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Hydroxysafflor yellow A, MedChemExpress
MedChemExpress Hydroxysafflor yellow A is a flavonoid derived and isolated from traditional Chinese medicine Carthamus tinctorius L.