CAS RN 7790-76-3
IUPAC Nombre:
dicalcium (phosphonatooxy)phosphonate
Sinónimos:
calcium pyrophosphate ca2P2O7 diphosphoric acid, calcium salt (2:1) calcium diphosphate
Peso molecular (g/mol):
254.1
Fórmula molecular:
Ca2O7P2
InChi Key:
JUNWLZAGQLJVLR-UHFFFAOYSA-J
SMILES:
[Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O
1
–
1
de
1
resultados
1
Fosfato de calcio (piro), 96 % mín., Thermo Scientific Chemicals
CAS: 7790-76-3 Fórmula molecular: Ca2O7P2 Peso molecular (g/mol): 254.10 Número MDL: MFCD00015983 Clave InChI: JUNWLZAGQLJVLR-UHFFFAOYSA-J Sinónimo: calcium pyrophosphate,calcium diphosphate,dicalcium diphosphate,unii-x69nu20d19,dicalcium pyrophosphate,alpha-calcium pyrophosphate,calcium phosphate ca2p2o7,diphosphoric acid, calcium salt,calcium pyrophosphate ca2p2o7,diphosphoric acid, calcium salt 1:2 PubChem CID: 24632 ChEBI: CHEBI:32598 Nombre IUPAC: dicalcio;fosfato fosfonato SMILES: [Ca++].[Ca++].[O-]P([O-])(=O)OP([O-])([O-])=O