CAS RN 77422-99-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PD 90780, MedChemExpress
MedChemExpress PD 90780 is a non peptide antagonist of nerve growth factor (NGF) binding to the P75 NGF receptor, inhibits NGF-p75 NTR interaction with IC50s values of 23.1 μM and 1.8 μM in PC12 cells and PC12 nnr5 cells, respectively .