CAS RN 7733-96-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HC-056456, MedChemExpress
MedChemExpress HC-056456 is an effective but not perfectly-selective blocker of CatSper channels. The [Na+]i rise is slowed by HC-056456 (IC50∼3 μM).