CAS RN 76494-51-4
IUPAC Nombre:
tetramethylpyrazine hydrochloride
Sinónimos:
tetramethylpyrazine hydrochloride
Peso molecular (g/mol):
172.66
Fórmula molecular:
C8H13ClN2
InChi Key:
RQKFOGXUTRDQPB-UHFFFAOYSA-N
SMILES:
Cl.CC1=NC(C)=C(C)N=C1C
1
–
2
de
2
resultados
1
Thermo Scientific Chemicals Hidrocloruro de tetrametilpirazina
CAS: 76494-51-4 Fórmula molecular: C8H12N2·HCl
2
Ligustrazine hydrochloride, MedChemExpress
MedChemExpress Ligustrazine (hydrochloride) is a natural product.