CAS RN 76252-06-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Nicainoprol, MedChemExpress
MedChemExpress Nicainoprol is a fast-sodium-channel blocking drug, which is a potent antiarrhythmic agent.