CAS RN 739366-20-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Anagliptina, TRC
CAS: 739366-20-2 Fórmula molecular: C19H25N7O2 Sinónimo: N-[2-[[2-[(2S)-2-Cyano-1-pirrolidinilo]-2-oxoetil]amino]-2-metilpropil]-2-metilpirazolo[1,5-a]pirimidina-6-carboxamida∣N-[2-([2-[(2S)-2-Cianopirrolidina-1-il]-2-oxoetil]amino)-2-metilpropilo-2-metilpirazolo[1,5-a]pirimidina-6-carboxamida∣Suiny
2
Anagliptin, MedChemExpress
MedChemExpress Anagliptin (SK-0403) is a highly selective, potent, orally active inhibitor of dipeptidyl peptidase 4 (DPP-4), with an IC50 of 3.8 nM, and less selective at DPP-8 and DDP-9 with IC50s of 68 nM and 60 nM, respectively.