CAS RN 73030-71-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Atractylenolide III, MedChemExpress
MedChemExpress Atractylenolide III is a major component of Atractylodes rhizome can induce apoptosis of the lung carcinoma cells.