CAS RN 72882-78-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PF-9366, MedChemExpress
MedChemExpress PF-9366 is a human methionine adenosyltransferase 2A (Mat2A) inhibitor, with an IC50 of 420 nM and a Kd of 170 nM.