CAS RN 725247-18-7
IUPAC Nombre:
(2E)-3-(4-{[2,4-bis(trifluoromethyl)phenyl]methoxy}-3-methoxyphenyl)-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide
Sinónimos:
(2E)-3-(4-{[2,4-bis(trifluoromethyl)phenyl]methoxy}-3-methoxyphenyl)-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide
Peso molecular (g/mol):
596.43
Fórmula molecular:
C23H13F9N4O3S
InChi Key:
HQFNFOOGGLSBBT-AWNIVKPZSA-N
SMILES:
COC1=CC(\C=C(/C#N)C(=O)NC2=NN=C(S2)C(F)(F)F)=CC=C1OCC1=CC=C(C=C1C(F)(F)F)C(F)(F)F
1
–
2
de
2
resultados
1
Tocris Bioscience™ XCT 790
CAS: 725247-18-7 Fórmula molecular: C23H13F9N4O3S
2
XCT790, MedChemExpress
MedChemExpress XCT-790 is a potent and selective inverse agonist for ERRα with an IC50 value of 0.37 μM. XCT-790 induces cell death in chemotherapeutic resistant cancer cells. XCT-790 (Compound 12) is inactive against ERRγ and the estrogen receptors ERα and ERβ.