CAS RN 72203-93-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Calcitetrol, MedChemExpress
MedChemExpress Calcitetrol(1(alpha), 24, 25-Trihydroxy VD3) is the hormonally active form of vitamin D with three hydroxyl groups.