CAS RN 72122-62-4
IUPAC Nombre:
(2S,3S)-2-{[(2S)-1-(2-{[(2S)-1-[(2S)-2-{[(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl]formamido}-3-phenylpropanoyl]pyrrolidin-2-yl]formamido}acetyl)pyrrolidin-2-yl]formamido}-3-methylpentanoic acid
Sinónimos:
(2S,3S)-2-{[(2S)-1-(2-{[(2S)-1-[(2S)-2-{[(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]pyrrolidin-2-yl]formamido}-3-phenylpropanoyl]pyrrolidin-2-yl]formamido}acetyl)pyrrolidin-2-yl]formamido}-3-methylpentanoic acid
Peso molecular (g/mol):
789.93
Fórmula molecular:
C41H55N7O9
InChi Key:
RKYJTDSQXOMDAD-JKXTZXEVSA-N
SMILES:
CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)CNC(=O)[C@@H]1CCCN1C(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H]1CCCN1C(=O)[C@@H](N)CC1=CC=C(O)C=C1)C(O)=O
1
–
2
de
2
resultados
1
MedChemExpress β-Casomorphin, bovine
MedChemExpress β-Casomorphin, bovine (β-Casomorphin-7 (bovine) ) is a opioid peptide with an IC50 of 14 μM in an Opioid receptors binding assay.
2
MedChemExpress β-Casomorphin, bovine
MedChemExpress β-Casomorphin, bovine (β-Casomorphin-7 (bovine) ) is a opioid peptide with an IC50 of 14 μM in an Opioid receptors binding assay.