CAS RN 717824-30-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Vidofludimus, MedChemExpress
MedChemExpress Vidofludimus(4SC-101; SC12267) is a novel immunosuppressive drug that inhibits DHODH; inhibits IL-17 secretion in vitro independently of effects on lymphocyte proliferation.