CAS RN 697235-39-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Episilvestrol, MedChemExpress
MedChemExpress Episilvestrol is a derivative of silvestrol, isolated from the fruits and twigs of Aglaia silvestris, and is a specific eIF4A-targeting translation inhibitor, with antitumor activity.