CAS RN 697235-38-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Silvestrol, MedChemExpress
MedChemExpress Silvestrol is a eukaryotic translation initiation factor 4A (eIF4A) inhibitor isolated from the fruits and twigs of Aglaia foveolata. Silvestrol induces autophagy and caspase-mediated apoptosis.