CAS RN 693228-63-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CYC-116, MedChemExpress
MedChemExpress CYC-116 is a potent aurora A and aurora B inhibitor with Kis of 8 and 9 nM, respectively.