CAS RN 6921-64-8
IUPAC Nombre:
1-(2-hydroxy-4-methylphenyl)ethan-1-one
Sinónimos:
1-(2-hydroxy-4-methylphenyl)ethanone
Peso molecular (g/mol):
150.18
Fórmula molecular:
C9H10O2
InChi Key:
LYKDOWJROLHYOT-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=C(C)C=C1O
1
–
2
de
2
resultados
1
2'-Hydroxy-4'-methylacetophenone, MedChemExpress
MedChemExpress 2'-Hydroxy-4'-methylacetophenone, a phenolic compound isolated from Angelicae koreana roots possesses acaricidal property. It could be used in the preparation of 4’-methyl-2’-[(p-tolylsulfonyl) oxy] acetophenone.