CAS RN 6909-62-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
N,N-Dimetilaminoetanol Fosfato (~70%), TRC
CAS: 6909-62-2 Fórmula molecular: C4 H12 N O4 P Peso molecular (g/mol): 169.12 Sinónimo: Phosphoric Acid Mono[2-(dimethylamino)ethyl] Ester,2-(Dimethylamino)ethanol Dihydrogen Phosphate,2-(Dimethylamino)ethyl Dihydrogen Phosphate,Demanyl Phosphate,N,N-Dimethylaminoethanol Monophosphate,N,N-Dimethylethanolamine Phosphate,P-DMEA,Pancar,Panclar,Phosphoryldimethylcolamine,Phosphoryldimethylethanolamine Nombre IUPAC: 2-(dimetilamino)etil dihidrógeno fosfato SMILES: CN(C)CCOP(=O)(O)O