CAS RN 690270-29-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Balapiravir
MedChemExpress Balapiravir (Ro 4588161; R1626) is an orally active prodrug of a nucleoside analogue inhibitor of the RNA-dependent RNA polymerase (RdRp) of HCV (R1479; 4'-Azidocytidine). Balapiravir has anti-HCV activity.
2
MedChemExpress Balapiravir
MedChemExpress Balapiravir (Ro 4588161; R1626) is an orally active prodrug of a nucleoside analogue inhibitor of the RNA-dependent RNA polymerase (RdRp) of HCV (R1479; 4'-Azidocytidine). Balapiravir has anti-HCV activity.
3
MedChemExpress Balapiravir
MedChemExpress Balapiravir (Ro 4588161; R1626) is an orally active prodrug of a nucleoside analogue inhibitor of the RNA-dependent RNA polymerase (RdRp) of HCV (R1479; 4'-Azidocytidine). Balapiravir has anti-HCV activity.