CAS RN 68252-28-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(R)-Oxiracetam, MedChemExpress
MedChemExpress (R)-Oxiracetam is the (R)-enantiomer of the nootropic drug oxiracetam. Oxiracetam (ISF 2522) is a nootropic drug of the racetam family and stimulant.