CAS RN 677338-12-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ PIK 90
Potent PI3K inhibitor, also used in hPSCs mesendoderm differentiation
2
PIK-90, MedChemExpress
MedChemExpress PIK-90 is a DNA-PK and PI3K inhibitor, which inhibits p110α, p110γ and DNA-PK with IC50s of 11, 18 and 13 nM, respectively.