CAS RN 675103-36-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ BAY 678
Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor
2
BAY-678, MedChemExpress
MedChemExpress BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).
3
BAY 678, Tocris Bioscience™
Potent and selective cell-permeable human neutrophil elastase (HNE) inhibitor