CAS RN 672926-33-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Kif15-IN-2, MedChemExpress
MedChemExpress Kif15-IN-2 is an inhibitor of the mitotic kinesin Kif15, and is used for the research of cellular proliferative diseases.