CAS RN 667463-85-6
IUPAC Nombre:
{6'-bromo-2'-oxo-1',2'-dihydro-[2,3'-biindolyliden]-3-yl}amino acetate
Sinónimos:
{6'-bromo-2'-oxo-1'H-[2,3'-biindolyliden]-3-yl}amino acetate
Peso molecular (g/mol):
398.22
Fórmula molecular:
C18H12BrN3O3
InChi Key:
ORWSYHWTBSHFIV-UHFFFAOYSA-N
SMILES:
CC(=O)ONC1=C2C=CC=CC2=NC1=C1C(=O)NC2=C1C=CC(Br)=C2
1
–
1
de
1
resultados
1
BIO-acetoxime, MedChemExpress
MedChemExpress BIO-acetoxime (BIA) is a potent and selective GSK-3 inhibitor, with IC50s of both 10 nM for GSK-3α/β. BIO-acetoxime has anticonvulsant and anti-infection activity.