CAS RN 66515-29-5
IUPAC Nombre:
2-amino-2-methyl-3-(phosphonooxy)propanoic acid
Sinónimos:
2-amino-2-methyl-3-(phosphonooxy)propanoic acid
Peso molecular (g/mol):
199.1
Fórmula molecular:
C4H10NO6P
InChi Key:
GSFCOAGADOGIGE-UHFFFAOYNA-N
SMILES:
CC(N)(COP(O)(O)=O)C(O)=O
1
–
1
de
1
resultados
1
MSOP, MedChemExpress
MedChemExpress MSOP is a selective group III metabotropic glutamate receptor antagonist with apparent KD of 51 μM for the L-AP4-sensitive presynaptic mGluR.