CAS RN 664969-54-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LF3, MedChemExpress
MedChemExpress LF3 is an antagonist of the β-Catenin/TCF4 interaction with antitumor activity; has an IC50 of 1.65 μM.