CAS RN 664338-39-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Arterolane, MedChemExpress
MedChemExpress Arterolane is an antimalarial agent, with IC50 of both 1.1 nM against P. falciparum Ro73 and W2, respectively.