CAS RN 65836-72-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DKM 2-93, MedChemExpress
MedChemExpress DKM 2-93 is a relatively selective inhibitor of UBA5 with an IC50 of 430 μM.