CAS RN 658084-23-2
IUPAC Nombre:
N-(3-chlorophenyl)-3-{[3,5-dimethyl-4-(4-methylpiperazine-1-carbonyl)-1H-pyrrol-2-yl]methylidene}-N-methyl-2-oxo-2,3-dihydro-1H-indole-5-sulfonamide
Sinónimos:
N-(3-chlorophenyl)-3-{[3,5-dimethyl-4-(4-methylpiperazine-1-carbonyl)-1H-pyrrol-2-yl]methylidene}-N-methyl-2-oxo-1H-indole-5-sulfonamide
Peso molecular (g/mol):
568.09
Fórmula molecular:
C28H30ClN5O4S
InChi Key:
FPYJSJDOHRDAMT-UHFFFAOYSA-N
SMILES:
CN(C1=CC(Cl)=CC=C1)S(=O)(=O)C1=CC2=C(NC(=O)C2=CC2=C(C)C(C(=O)N3CCN(C)CC3)=C(C)N2)C=C1
1
–
2
de
2
resultados
1
SU11274, MedChemExpress
MedChemExpress SU11274 is a selective Met inhibitor with IC50 of 10 nM, but has no effects on PGDFRβ, EGFR or Tie2.
2