CAS RN 65550-32-5
IUPAC Nombre:
ethyl 2-methylpyridine-4-carboxylate hydrochloride
Sinónimos:
ethyl 2-methylpyridine-4-carboxylate hydrochloride
Peso molecular (g/mol):
201.65
Fórmula molecular:
C9H12ClNO2
InChi Key:
DPRMHHCAVHGENN-UHFFFAOYSA-N
SMILES:
Cl.CCOC(=O)C1=CC(C)=NC=C1
1
–
1
de
1
resultados
1
Clorhidrato 2-metilpiridina-4-carboxilato de etilo, 95 %, Thermo Scientific Chemicals
CAS: 65550-32-5 Fórmula molecular: C9H12ClNO2 Peso molecular (g/mol): 201.65 Número MDL: MFCD09991690 Clave InChI: DPRMHHCAVHGENN-UHFFFAOYSA-N Sinónimo: ethyl 2-methylisonicotinate hydrochloride,ethyl 2-methylpyridine-4-carboxylate hydrochloride,ethyl 2-methylisonicotinate hcl,2-methylisonicotinic acid ethyl ester hydrochloride,acmc-20al0t,2-methyl-isonicotinic acid ethyl ester hydrochloride,2-methyl-4-pyridinecarboxylic acid ethyl ester hydrochloride,ethyl 2-methylpyridine-4-carboxylate-hydrogen chloride 1/1,4-pyridinecarboxylic acid, 2-methyl-, ethyl ester, hydrochloride PubChem CID: 53229930 SMILES: Cl.CCOC(=O)C1=CC(C)=NC=C1