CAS RN 6501-72-0
IUPAC Nombre:
2-(3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid
Sinónimos:
(3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid
Peso molecular (g/mol):
205.21
Fórmula molecular:
C11H11NO3
InChi Key:
MUFJHYRCIHHATF-UHFFFAOYNA-N
SMILES:
OC(=O)CC1CC(=NO1)C1=CC=CC=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ GIT 27
CAS: 6501-72-0 Fórmula molecular: C11H11NO3
2
VGX-1027, MedChemExpress
MedChemExpress VGX-1027 is an orally active isoxazole compound that exhibits various immunomodulatory properties. VGX-1027 targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, IL-10. VGX-1027 has antidiabetogenic effects by limiting cytokine-mediated immunoinflammatory events.