CAS RN 64134-30-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Hexa-His
MedChemExpress Hexa-His is a peptide consisting of 6 His residues, used as a metal binding site for the recombinant protein.
2
MedChemExpress Hexa-His
MedChemExpress Hexa-His is a peptide consisting of 6 His residues, used as a metal binding site for the recombinant protein.
3
MedChemExpress Hexa-His
MedChemExpress Hexa-His is a peptide consisting of 6 His residues, used as a metal binding site for the recombinant protein.