CAS RN 64111-99-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress HAE
MedChemExpress HAE is a 3-amino acid peptide which consists of histidine, alanine and glutamate.
2
MedChemExpress HAE
MedChemExpress HAE is a 3-amino acid peptide which consists of histidine, alanine and glutamate.
3
MedChemExpress HAE
MedChemExpress HAE is a 3-amino acid peptide which consists of histidine, alanine and glutamate.