CAS RN 63238-66-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Benzoylhypaconine, MedChemExpress
MedChemExpress Benzoylhypaconine (Benzoylhypacoitine) is a monoester Aconitum alkaloid, is the main pharmacologic and toxic component.