CAS RN 629664-81-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Turofexorate isopropyl, MedChemExpress
MedChemExpress Turofexorate isopropyl (FXR-450) is a potent, selective, and orally bioavailable FXR agonist with EC50 of 4 nM.